Learn More
BEZWADA BIOMEDICAL LLC Poly(caprolactone-co-L-lactide) caprolactone:L-lactide 90:10, viscosity 1.0 to 2.0 dl/g, 5.0 GM
SDP
Supplier:  BEZWADA BIOMEDICAL LLC 1391
Encompass_Preferred
The presence of lactide repeat units in the copolymer reduces the degree of close packing between caprolactone repeat units leaving the polymer with nice elastic properties while maintaining a relatively slow degradation profile. This allows PCL-PLA copolymers to be used in drug delivery (liquid polymers) and medical device coatings.
Catalog No. 50-190-4459