Learn More
LUNA NANOTECH INC Pcr Plate Magnetic Rack
Supplier:  LUNA NANOTECH INC MGRPCRP
Encompass_Preferred
96-well PCR Plate Magnetic Separation Rack
- Designed to be used with 0.2 mL 96-well PCR plates, PCR tubes, 8-strip PCR tubes
- 24 magnets are positiont between the wells of the plates for efficent magnetic separation
- Can be used for high throughput separation of up to 96 samples
- Housing material: PMMA/Acrylic
- Magnet: N38 Neodymium (NdFeB)
Catalog No. 50-256-6966