Learn More
LUNA NANOTECH INC 96-Well Pcr Product Purification Kit, 384 Preps
Supplier:  LUNA NANOTECH INC NKP105384
Encompass_Preferred
PuroSPIN-96™ PCR Product Purification Kit, 96-Well Plate Format, 384 Preps
The PuroSPIN™ PCR Product Purification 96 Kit is designed to provide a simple and easy-to-use high-throughput method for purification of PCR DNA products (100 bp − 10 kb) from enzymatic reactions.
This purification technology is based on nucleic acids binding to silica purification plates, which allows for efficient removal of protein, salt, and other contaminants. Silica purification plates provide a faster, simpler, and safer alternative to traditional nucleic acid purification techniques such as phenol-chloroform extraction.
The PuroSPIN™ buffer formulations produce highly purified DNA product for downstream applications such as sequencing, ligation, transfection, Southern blotting, qPCR, or microarray analysis.
Features:
- High throughput sample processing using 96-well silica membrane purification plates
- Efficient removal
Catalog No. 50-256-6920