Learn More
Medchemexpress LLC PCS1055 | 357173-55-8 | 98.0% | 5 MG
SDP
Supplier:  Medchemexpress LLC HY122203A5MG
Encompass_Preferred
PCS1055 is a selective and competitive antagonist of the muscarinic M4 receptor (mAChR4) with low-nanomolar potency and additional cholinesterase inhibitory activity. It is intended for research use in receptor pharmacology and enzymology studies.
  • Selective mAChR4 antagonist with IC50 18.1 nM.
  • High affinity in radioligand binding (Ki 6.5 nM; Kd 5.72 nM).
  • Cholinesterase inhibitor activity (electric eel AChE IC50 22 nM; human AChE IC50 120 nM).
  • Molecular formula C27H32N4; molecular weight 412.57 g/mol.
  • Supplied in small research quantities with recommended storage conditions for powder and solutions.
Catalog No. 50-002-30723