Learn More
Cayman Chemical PCSK9-DyLIght633 50ug
SDP
Supplier:  Cayman Chemical 3059250 UG
Encompass_Preferred
Source: Active recombinant C-terminal His-tagged enzyme expressed in HEK293 cells. Conjugated to DyLight™ 633 • Amino acids: 1-692 (full length) • MW: 13.8 kDa prodomain + 59 kDa mature form
Catalog No. 50-297-8801