Learn More
Medchemexpress LLC PCSK9 Human Biotin 10ug | 10ug
SDP
Supplier:  Medchemexpress LLC HYP7882410UG
Encompass_Preferred
The PCSK9 protein is an important regulator of plasma cholesterol homeostasis affecting LDL receptor family members such as LDLR and VLDLR It promotes their intracellular degradation and enhances hepatic LDLR degradation through non-proteolytic mechanisms PCSK9 Protein Human (Biotinylated HEK293 D374Y His-Avi) is the recombinant human-derived PCSK9 protein expressed by HEK293 with C-Avi C-His labeled tag and D374Y mutation
Catalog No. 50-001-94911