Learn More
Genscript Corporation PCT 18D2 mAb Mouse
Supplier:  Genscript Corporation V01803100
Encompass
PCT a 116 amino acid (aa) protein is comprised of three sections including a 57 aa N-terminal PCT a 32 aa calcitonin and a 21 aa katacalcin Calcitonin is a hormone derived from PCT cleavage PCT is a good diagnosis marker for bacterial infection Other diseases such as sepsis inflammation surgery heat shock burn injuries and cardiogenic shock can also cause an increase of PCT level in blood
Catalog No. 50-000-57693
Only null left
Add to Cart
Request a Quote