Learn More
System Biosciences LLC Exosome Cyto-Tracer, pCT-CD9-GFPpCT-CD9-GFP (pCMV, Exosome/Secretory, CD9 Tetraspanin Tag) = 10ug
SDP
Supplier:  System Biosciences LLC CYTO122PA1
Encompass_Preferred
Label exosomes with GFP by expressing the pCT-CD9-GFP Cyto-Tracer in the parent cells. Great for tracking exosomes and monitoring cargo delivery. Monitor exosome dynamics with the pCT-CD9-GFP (pCMV) Exosome Cyto-Tracer (also available as a CD9-RFP fusion, pCT-CD9-RFP). Provided as either lentivector or pre-packaged virus, this Cyto-Tracer fuses GFP to the tetraspanin CD9, which then gets packaged into exosomes produced by the transfected cells. Great for creating stable cell lines that produce GFP-labeled exosomes as well as labeling exosomes from primary cells, tumor cells, stem cells, and more.
Catalog No. NC3244672