Learn More
System Biosciences LLC Exosome Cyto-Tracer, pCT-CD9-RFPpCT-CD9-RFP (pCMV, Exosome/Secretory, CD9 Tetraspanin Tag) = 10ug
Supplier: System Biosciences LLC CYTO123PA1
Label exosomes with RFP by expressing the pCT-CD9-RFP Cyto-Tracer in the parent cells. Great for tracking exosomes and monitoring cargo delivery. Monitor exosome dynamics with the pCT-CD9-RFP (pCMV) Exosome Cyto-Tracer (also available as a CD9-GFP fusion, pCT-CD9-GFP). Provided as either lentivector or pre-packaged virus, this Cyto-Tracer fuses RFP to the tetraspanin CD9, which then gets packaged into exosomes produced by the transfected cells. Great for creating stable cell lines that produce RFP-labeled exosomes as well as labeling exosomes from primary cells, tumor cells, stem cells, and more.