Learn More
Selleck Chemical LLC Palbociclib (PD-0332991) HCl 50mg 827022-32-2
Supplier: Selleck Chemical LLC S111650MG
Palbociclib (PD-0332991) HCl is a highly selective inhibitor of CDK4/6 with IC50 of 11 nM/16 nM in cell-free assays, respectively. It shows no activity against CDK1/2/5, EGFR, FGFR, PDGFR, InsR, etc. Phase 3. *For Research & Development use only. Product is not intended for drug, household, or human consumption.