Learn More
Medchemexpress LLC PD-1 Cynomolgus 14 5ug
SDP
Supplier:  Medchemexpress LLC HYP706765UG
Encompass_Preferred
Recombinant cynomolgus PD-1 extracellular domain (P21-Q167), expressed in HEK293 cells and supplied as a His-tagged, lyophilized protein. The product is formulated from PBS (pH 7.4), has low endotoxin, and is functionally validated by ELISA for binding to PD-L1 (ED50 260.2 ng/mL). Recommended storage is -20°C; reconstituted material is stable short term at 4°C.
Catalog No. 50-001-84427