Learn More
Medchemexpress LLC PD-1/PD-L1-in-23 | 2597056-04-5 | C32H30BrCl2N3O6 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14577410MG
Encompass_Preferred
PD-1/PD-L1-IN-23 is a small-molecule research compound that inhibits the PD-1/PD-L1 immune checkpoint pathway. Supplied as a solid powder in milligram-scale packaging, it is intended for in vitro and preclinical studies to probe PD-1/PD-L1 interactions in biochemical and cellular assays.
  • High purity (>98%).
  • Chemical formula C32H30BrCl2N3O6; molecular weight 703.41 g/mol.
  • Supplied as a solid powder suitable for dissolution in common laboratory solvents.
  • Intended for in vitro and preclinical research use; not for clinical or human use.
  • Useful for studies of PD-1/PD-L1 interaction and checkpoint inhibition assays.
Catalog No. 50-002-58793