Learn More
Apexbio Technology LLC PD-1/PD-L1 inhibitor 1(Synonyms: BMS-202, PD-1/PD-L1 interaction inhibitor 1, PD-L1 inhibitor BMS-202, BMS202), 25mg, CAS: 1675201-83-8.
SDP
Supplier:  Apexbio Technology LLC B617225
Encompass_Preferred
PD-1/PD-L1 inhibitor 1 (CAS 1675201-83-8) is a small molecule inhibitor targeting the interaction between programmed cell death protein 1 (PD-1) and its ligand PD-L1 By disrupting PD-1/PD-L1 signaling this agent restores and enhances T-cell activation demonstrating immunomodulatory properties In vitro assays (HTRF binding assay) report IC50 values ranging from 6 to 100 nM Current preclinical evidence positions PD-1/PD-L1 inhibitor 1 as a candidate compound for studying tumor immunity and infectious diseases such as hepatitis C virus infection
Catalog No. 50-000-60479