Learn More
Apexbio Technology LLC PD-1/PD-L1 inhibitor 2(Synonyms: BMS-202, BMS202, PD-L1 inhibitor BMS-202, PD-1/PD-L1 antagonist BMS-202), 5mg, CAS: 1675203-84-5.
SDP
Supplier:  Apexbio Technology LLC B60235
Encompass_Preferred
PD-1/PD-L1 inhibitor 2 (CAS 1675203-84-5) also known as BMS-202 is a small molecule inhibitor targeting the PD-1/PD-L1 immune checkpoint interaction It binds directly to human PD-L1 protein disrupting its interaction with PD-1 receptor displaying an IC50 of 18 nM and a binding dissociation constant (KD) of approximately 8 M By obstructing the PD-1/PD-L1 signaling axis this compound is valuable in studies investigating tumor-associated immune modulation and understanding PD-L1 s biological function within the tumor immune microenvironment
Catalog No. 50-000-60426