Learn More
TARGETMOL CHEMICALS INC PD 128042 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2266510MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. PD 128042 (CI 976) is a potent, orally active, and selective ACAT inhibitor (IC50 73 nM) as well as a lysophospholipid acyltransferase (LPAT) inhibitor. CI 976 inhibits Golgi-associated LPAT activity (IC50 15 uM) and multiple membrane trafficking steps. Purity 99.44%
Catalog No. 50-287-7735