Learn More
Medchemexpress LLC PD 128907 hydrochloride | 112960-16-4 | MFCD00210210 | 99.5% | 285.77 | C14H20ClNO3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY100539A10MG
Encompass_Preferred
PD 128907 hydrochloride is the hydrochloride salt of a potent and selective dopamine D3 receptor agonist intended for in vitro pharmacology and receptor-binding research. The material is supplied with a high stated purity and defined chemical characterization suitable for laboratory assays.
  • Selective dopamine D3 receptor agonist with reported EC50 of 0.64 nM.
  • High purity (99.51%).
  • Molecular weight 285.77 and molecular formula C14H20ClNO3.
  • Suitable for in vitro receptor binding and functional assays.
  • Available in multiple small-scale milligram pack sizes for laboratory use.
Catalog No. 50-002-14980