Learn More
Medchemexpress LLC (+)-PD 128907 hydrochloride | 300576-59-4 | 98.92% | 285.77 | 50 MG
Supplier: Medchemexpress LLC HY11000010MG
(+)-PD 128907 hydrochloride is a selective dopamine D2/D3 receptor agonist, with Kis of 1.7 nM and 0.84 nM for human and rat D3 receptors, and 179 nM and 770 nM for human and rat D2 receptors, respectively. This product is for research use only and is not sold to patients.
- Selective dopamine D2/D3 receptor agonist.
- Kis of 1.7 nM (human D3) and 0.84 nM (rat D3).
- Selectivity over human and rat dopamine D2 receptor (>100-fold and 900-fold, respectively).
- Decreases dialysate DA levels in D3 knock out mice and wild-type mice, with higher potency in wild-type.