Learn More
Enzo Life Sciences PD-146176 (10mg). CAS: 4079-26-9
SDP
Supplier:  Enzo Life Sciences BMLEI3560010
Encompass_Preferred
A potent and selective 15-lipoxygenase inhibitor which lacks significant nonspecific antioxidant properties. It suppresses atherogenesis and limits atherosclerotic lesion development in the rabbit. It reduces oxidant stress-induced apoptosis in endothelial cells and inhibits proliferation in 15-LO overexpressing PC3 cells. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (50mg/mL) and ethanol (10mg/mL). Long Term Storage: Ambient.
Catalog No. 50-201-0741