Learn More
AdipoGen PD 150,606 (25 mg)
SDP
Supplier:  AdipoGen AGCR10066M025
Encompass_Preferred
Chemical. CAS: 426821-41-2. Formula: C9H7IO2S. MW: 306.1. Cell permeable and selective calpain-1 and -2 inhibitor. Interacts with calcium-binding domain of calpain. Neuroprotective. Ca2+-permeable AMPA receptor inhibitor. Apoptosis inhibitor Autophagy activator by preventing cleavage of ATG5
Catalog No. 50-205-3821