Learn More
Selleck Chemical LLC PD 151746
SDP
Supplier:  Selleck Chemical LLC S742425MG
Encompass_Preferred
PD 151746 is a selective cell-permeable calpain inhibitor with Ki of 0 26 M for -Calpain about 20-fold selectivity over m-calpain
Catalog No. 50-000-01458