Learn More
Apexbio Technology LLC PD 161570 192705-80-9 10mg
SDP
Supplier:  Apexbio Technology LLC B748610
Encompass_Preferred
PD 161570 (CAS 192705-80-9) is a small-molecule inhibitor that acts through ATP-competitive binding to suppress the phosphorylation of human FGF-1 receptors In addition to targeting FGF-1R PD 161570 exhibits inhibitory activity against several other tyrosine kinases including PDGFR EGFR and c-Src The compound also interferes with signaling pathways associated with BMP and TGF- Due to its capacity to modulate multiple kinase-driven processes PD 161570 serves as a valuable tool in studies of cell signaling pathways and cellular proliferation mechanisms
Catalog No. 50-000-88237