Learn More
Abcam PD 168393, EGFR inhibitor, 1MG
Supplier:  Abcam AB1451871MG
Encompass_Preferred
MW 369.2 Da, Purity >98%. Potent, irreversible, cell-permeable, selective EGFR inhibitor (IC₅₀ values are 0.7, 0.08, 5.7, 12 and 15 nM for EGF receptor tyrosine kinase, erbB1, erbB2, erbB4 and SK-BR-3, respectively). Binds to the catalytic domain. Inhibits cancer cell growth. Blocks reactive astrogliosis and shows neuroprotective effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5420