Learn More
Abcam PD 168568 dihydrochloride, D4 antagonist, 10MG
Supplier:  Abcam AB12061110MG
Encompass_Preferred
MW 422.4 g/mol, Purity >99%. Potent and selective D₄ antagonist (Ki = 8.8 nM at D₄, and 1842 nM at D₂). Inhibits amphetamine stimulated locomotor activity in vivo; orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7427