Learn More
Medchemexpress LLC HY-10578 10mg Medchemexpress, PD 169316 CAS:152121-53-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1057810MG
Encompass_Preferred
Medchemexpress, HY-10578 10mg PD 169316 CAS:152121-53-4 PD 169316 is a potent, cell-permeable and selective p38 MAP kinase inhibitor, with IC50 of 89 nM. PD169316 selectively inhibits the kinase activity of the phosphorylated p38 without hindering upstream kinases to phosphorylate p38. PD169316 shows antiviral activity against Enterovirus71. PD169316 shows antiviral activity against Enterovirus71. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8293