Learn More
Enzo Life Sciences PD 173074 (5mg). CAS: 219580-11-7
SDP
Supplier:  Enzo Life Sciences ENZCHM1120005
Encompass_Preferred
A cell-permeable compound that acts as a potent, ATP-competitive, and reversible inhibitor of FGF and VEGF receptors. PD 173074 inhibits PDGFR and c-Src only at much higher concentration. Shown to inhibit the autophosphorylation of endogenous FGFR1 and overexpressed VEGFR2. Alternative name: 1-t-Butyl-3-(6-(3,5-dimethoxyphenyl)-2-(4-diethylaminobutylamino)-pyrido[2,3-d]pyrimidin-7-yl)urea. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO or methanol. Long Term Storage: -80°C.
Catalog No. 50-201-3389