Learn More
Sigma Aldrich Fine Chemicals Biosciences Pd EnCatR 30 extent of lab10G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 64471410G
Encompass
It is an encapsulated catalyst (EnCat(R)) wherein palladium acetate is microencapsulated within a polyurea matrix which prevents palladium leaching.
Catalog No. 50-290-7740