Learn More
Genetex Inc PD-L1 Monoclonal Antibody [H302] HistoMAX™ (GTX639925) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX639925
Encompass_Preferred
The PD-L1 antibody [H302] HistoMAX™ (GTX639925) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human oral cavity squamous cell carcinoma (OCSCC) tissue using GTX639925 PD-L1 antibody [H302] HistoMAX. Strong predominantly membranous PD-L1 positivity of all tumor cells of a squamous cell carcinoma of the oral cavity. Some inflammatory cells are also PD-L1 positive.
Catalog No. NC3669085