Learn More
Genetex Inc PD-L1 Monoclonal Antibody [MSVA-711R] HistoMAX™ (GTX04362) GeneTex - IHC-Optimized Antibody
SDP
Supplier:  Genetex Inc GTX04362
Encompass_Preferred
"The PD-L1 antibody [MSVA-711R] HistoMAX™ (GTX04362) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human invasive breast cancer of no special type (NST) tissue using GTX04362 PD-L1 antibody [MSVA-711R] HistoMAX. Breast cancer NST with PD-L1 negative tumor cells but intense PD-L1 staining in tumor associated inflammatory cells."
Catalog No. NC3660474