Learn More
Medchemexpress LLC PD-L1 Cynomolgus/Rh 100ug | 100ug
SDP
Supplier:  Medchemexpress LLC HYP77347100UG
Encompass_Preferred
The PD-L1 protein critically regulates immune tolerance by acting as a ligand for PDCD1/PD-1 regulating T cell activation threshold and limiting effector responses It may act as a costimulatory molecule for IL10-producing T cell subsets PD-L1 Protein Cynomolgus/Rhesus Macaque (HEK293 His) is the recombinant Rhesus Macaque-derived PD-L1 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-86840