Learn More
Medchemexpress LLC PD-L1 inhibitory peptide TFA | 1983.29 | 50MG
SDP
Supplier:  Medchemexpress LLC HYP10950A50MG
Encompass_Preferred
PD-L1 inhibitory peptide TFA is an inhibitor peptide targeting programmed cell death ligand 1 (PD-L1). It binds to PD-L1, relieving immunosuppression and restoring the antitumor activity of T cells, making it promising for cancer research.
  • Targets programmed cell death ligand 1 (PD-L1)
  • Relieves immunosuppression
  • Restores antitumor activity of T cells
  • Promising for cancer research
Catalog No. 50-004-22422