Learn More
Sigma Aldrich Fine Chemicals Biosciences PD MultiTrapTM G 25 Cytiva
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE28918006
Encompass
PD MultiTrap(TM) G-25 is designed for high-throughput desalting and buffer exchange of multiple samples. Supplied in packs of 4 plates. Instructions included. Plates can be processed by vacuum or centrifugation. Suitable for manual use as well as for automation together with robotic systems. Sample preparation in the range of 70 to 130 mu without sample dilution. PD MultiTrap(TM) G-25 is prepacked with 500 mu Sephadex(R) G-25 Medium/well. The 96-well plates are manufactured from biocompatible polypropylene. Convenient and fast small-scale cleanup of proteins and other large biomolecules with high reproducibility well-to-well and plate-to-plate. High desalting capacity (85%) and recovery (70-90%).
Catalog No. 50-290-9057