Learn More
Medchemexpress LLC Pd(Xantphos)Cl2 | 205319-10-4 | >=98.0% | 755.94 | 5 G

Supplier: Medchemexpress LLC HYW0130665G
Pd(Xantphos)Cl2 is a palladium(II) complex catalyst used in organic synthesis research.
- Can be used in catalytic reactions for organic synthesis.
- Intended for research use only.
- Store at 4°C, protect from light, stored under nitrogen.
- Soluble in DMSO for in vitro applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.