Learn More
Apexbio Technology LLC PD123319,5mg CAS# 130663-39-7
SDP
Supplier:  Apexbio Technology LLC B22065
Encompass_Preferred
PD123319 is a non-peptide inhibitor of angiotensin II receptor with IC50 value of 34nM [1].Angiotensin II play roles in a variety of physiological functions. Among these, the most prominent is vascular contraction. Unlike previous drugs act as inhibitors of the formation of Ang II or ACE, PD123319 is an antagonist of angiotensin II receptor. PD123319 shows inhibition potency in both rat adrenal and brain binding assay with IC50 values of 34nM and 210nM, respectively. It is found to prevent Ang II from binding the bovine zona glomerulosa microsomal preparation with IC50 value of 6.9nM in the binding assay using microsome. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0169