Learn More
Selleck Chemical LLC PD128907 HCl S2168-5mg
SDP
Supplier:  Selleck Chemical LLC S21685MG
Encompass_Preferred
PD 128907 HCl is a potent and selective dopamine D3 receptor agonist with EC50 of 0 64 nM exhibits 53-fold selectivity over dopamine D2 receptor
Catalog No. 50-311-2676