Learn More
Medchemexpress LLC HY-103157 5mg Medchemexpress, PD146176 CAS:4079-26-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1031575MG
Encompass_Preferred
Medchemexpress, HY-103157 5mg PD146176 CAS:4079-26-9 PD146176 (NSC168807) is a 15-Lipoxygenase (15-LO) inhibitor, which inhibits rabbit reticulocyte 15-LO with a K i of 197 nM. PD146176 (NSC168807) has a dramatic effect in reducing atherogenesis [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2237