Learn More
Selleck Chemical LLC PD153035-5mg
SDP
Supplier:  Selleck Chemical LLC S65465MG
Encompass_Preferred
PD153035 is a specific and potent inhibitor of the epidermal growth factor (EGF) receptor tyrosine kinase (Ki = 5.2 pM).
Catalog No. 50-238-9947