Learn More
Selleck Chemical LLC PD153035
SDP
Supplier:  Selleck Chemical LLC S654625MG
Encompass_Preferred
PD153035 is a specific and potent inhibitor of the epidermal growth factor (EGF) receptor tyrosine kinase (Ki 5 2 pM)
Catalog No. 50-000-02074