Learn More
Medchemexpress LLC HY-12013 5mg Medchemexpress, PD153035 (Hydrochloride) CAS:183322-45-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY120135MG
Encompass_Preferred
Medchemexpress, HY-12013 5mg PD153035 (HCl) CAS:183322-45-4 PD153035 Hydrochloride (SU-5271 Hydrochloride) is a potent EGFR inhibitor with Ki and IC50 of 6 and 25 pM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9582