Learn More
Medchemexpress LLC PD153035 hydrochloride | 183322-45-4 | ≥98.0% | 25 MG
SDP
Supplier:  Medchemexpress LLC HY1201325MG
Encompass_Preferred
PD153035 hydrochloride is a potent small-molecule epidermal growth factor receptor (EGFR) inhibitor used in biochemical and cell-based research to block EGFR signaling. It binds EGFR with high affinity (Ki ≈ 6 pM) and inhibits activity at low picomolar concentrations (IC50 ≈ 25 pM). The compound is supplied as the hydrochloride salt and is intended for research use.
  • Potent EGFR inhibition with Ki ≈ 6 pM and IC50 ≈ 25 pM.
  • Suitable for biochemical and cell-based assays.
  • Provided as a hydrochloride salt for improved solubility.
  • Molecular formula C16H15BrClN3O2; molecular weight 396.67 g/mol.
  • Store sealed, away from moisture; in solution store at -80°C for long-term stability.
Catalog No. 50-002-33430