Learn More
TARGETMOL CHEMICALS INC PD98059 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T262350MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. PD98059 is an MEK inhibitor that inhibits MEK1 and MEK2 (IC50=2/50 uM) and is non-ATP-competitive. PD98059 is also antagonistic as a ligand for AHR. PD98059 inhibits autophagy. Purity 98.56%
Catalog No. 50-287-7744