Learn More
Selleck Chemical LLC PD98059 10mg 167869-21-8
SDP
Supplier:  Selleck Chemical LLC S117710MG
Encompass_Preferred
PD98059 is a non-ATP competitive MEK inhibitor with IC50 of 2 ?M in a cell-free assay, specifically inhibits MEK-1-mediated activation of MAPK; does not directly inhibit ERK1 or ERK2. PD98059 is a ligand for the aryl hydrocarbon receptor (AHR) and functions as an AHR antagonist. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-888-2