Learn More
BEZWADA BIOMEDICAL LLC Poly(P-dioxanone-co-L-lactide), p-dioxanone:L-lactide 95:5, viscosity 1.0 to 2.0 dl/g, 5.0 GM
SDP
Supplier:  BEZWADA BIOMEDICAL LLC 63955
Encompass_Preferred
Copolymers of PDS with L-lactide is soft and flexible and has exellent pliability. Biomedical devices include surgical sutures, staples, meshes, and controlled drug delivery.BSR profile longer than PDS.Biomedical devices include surgical sutures, staples, meshes, and controlled drug delivery.
Catalog No. 50-190-4488