Learn More
Medchemexpress LLC Pdc/pkd-in-1 | 1071135-06-2 | 98.35% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1281421MG
Encompass_Preferred
PKC/PKD-IN-1 is an orally active dual protein kinase C/D (PKC/PKD) inhibitor with an IC50 value of 0.6 nM for PKD1. It can attenuate high-salt diet-induced cardiac hypertrophy and can be used in the study of heart failure. This product is for research use only and not for sale to patients.
  • Orally active dual protein kinase C/D (PKC/PKD) inhibitor
  • Has an IC50 value of 0.6 nM for PKD1
  • Attenuates high-salt diet-induced cardiac hypertrophy
  • Used in the study of heart failure
  • For research use only
Catalog No. 50-003-39210