Learn More
Aviva Systems Biology Corp PDCD8 Antibody - C-terminal region (ARP40199_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40199_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PDCD8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PDCD8. Alternate names: AIF, PDCD8, COXPD6ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-57335