Learn More
Medchemexpress LLC HY-108360 10mg Medchemexpress, PDD 00017273 CAS:1945950-21-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10836010MG
Encompass_Preferred
Medchemexpress, HY-108360 10mg PDD 00017273 CAS:1945950-21-9 PDD 00017273 is a potent inhibitor of Poly(ADP-ribose) Glycohydrolase ( PARG ), with an IC 50 of 26 nM, and a K D of 1.45 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2318