Learn More
Selleck Chemical LLC PDD00017273 S8862-100MG
SDP
Supplier:  Selleck Chemical LLC S8862100MG
Encompass_Preferred
PDD00017273 is a potent and selective poly (ADP ribose) glycohydrolase (PARG) inhibitor with an IC50 of 26 nM PDD00017273 exhibits 350-fold selectivity for PARG over a panel of ion channels enzymes and receptors including PARP1 and ARH3
Catalog No. 50-000-25033