Learn More
TARGETMOL CHEMICALS INC PDE4B-IN-2 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T368325MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PDE4B-IN-2 (A 33) is an orally active and selective PDE4B inhibitor with an IC50 of 15 nM. PDE4B-IN-2 inhibits PDE4D (IC50=1.7 µM). PDE4B-IN-2 exhibits potent anti-inflammatory effects. purity: 99%
Catalog No. 50-258-0165