Learn More
Medchemexpress LLC Pde7-in-3 | 908570-13-8 | 99.9% | 364.82 g/mol | C18H21ClN2O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10032625MG
Encompass_Preferred
PDE7-IN-3 is a selective phosphodiesterase 7 (PDE7) inhibitor with reported analgesic activity. It is supplied as a white to off-white solid and is used in vitro and in vivo research to study inflammatory, neuropathic, visceral, and nociceptive pain pathways.
  • Selective PDE7 inhibition for pharmacological studies.
  • High purity (99.94%) suitable for research applications.
  • Available in multiple small-scale quantities and DMSO solutions.
  • Identified by CAS number 908570-13-8 for unambiguous referencing.
  • Provided with datasheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-15064