Learn More
Medchemexpress LLC PDGF-AA Human His 2ug
SDP
Supplier:  Medchemexpress LLC HYP708252UG
Encompass_Preferred
Recombinant human PDGF-AA protein with an N-terminal 6xHis tag, supplied as a lyophilized powder. Expressed in E. coli, the protein is biologically active in fibroblast proliferation assays (ED50 ≈ 53.4 ng/mL) and is intended for research into cell proliferation, migration, and wound healing.
  • Expressed in E. coli.
  • Contains N-terminal 6xHis tag for detection and purification.
  • Lyophilized for long-term storage and convenient reconstitution.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Low endotoxin (<1 EU/μg) suitable for cell-based assays.
  • Demonstrated biological activity (ED50 ~53.4 ng/mL in NIH-3T3 assay).
  • Store lyophilized at -20°C; aliquot and freeze for extended storage.
Catalog No. 50-001-97012