Learn More
Medchemexpress LLC PDGF R beta Human 5ug | 5ug
SDP
Supplier:  Medchemexpress LLC HYP771235UG
Encompass_Preferred
PDGF R protein is integral in embryonic development and regulates cell proliferation survival differentiation chemotaxis migration and vascular development It crucially recruits pericytes and smooth muscle cells to the endothelium promoting vascular smooth muscle cell migration and neointimal formation at the site of injury PDGF R beta Protein Human (HEK293 C-His-Avi) is the recombinant human-derived PDGF R beta protein expressed by HEK293 with C-His C-Avi labeled tag
Catalog No. 50-001-85694